missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester, 95%, Thermo Scientific Chemicals

Product Code. 11496170
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11496170 1 g 1g
15487655 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496170 Supplier Thermo Scientific Alfa Aesar Supplier No. H27097.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165881-36-7
Molecular Formula C14H24BBrO2
Molecular Weight (g/mol) 315.058
MDL Number MFCD09037500
InChI Key DQGAIMVYOVPBSD-FMSGJZPZSA-N
Synonym 4-bromobutylboronic acid 1s,2s,3r,5s-+-2,3-pinanediol ester, 4-bromobutylboronate ester, 4-bromobutaneboronic acid 1s,2s,3r,5s-+-2,3-pinanediol ester, 1s,2s,3r,5s-+-2,3-pinanediol 4-bromobutylboronate ester, 1s,2s,6r,8s-4-4-bromobutyl-1,8-dihydrogenio-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.02,6 decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0?,? decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0^ 2,6 decane, 1s,2s,6r,8s-4-4-bromobutyl-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo 6.1.1.0_,? decane, 3as,4s,6s,7ar-2-4-bromobutyl-3a,5,5-trimethylhexahydro-2h-4,6-methano-1,3,2-benzodioxaborole, 3as,4s,6s,7ar-2-4-bromobutyl-3a,5,5-trimethylhexahydro-4,6-methanobenzo d 1,3,2 dioxaborole
PubChem CID 44784918
SMILES B1(OC2CC3CC(C3(C)C)C2(O1)C)CCCCBr

Specifications

Refractive Index 1.496
Quantity 1 g
Optical Rotation +18° (c=1 in Chloroform)
Formula Weight 315.06
Percent Purity 95%
Chemical Name or Material 4-Bromobutylboronic acid (1S,2S,3R,5S)-(+)-2,3-pinanediol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.