missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Bromobutyl)triphenylphosphonium Bromide, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | (4-Bromobutyl)triphenylphosphonium Bromide |
| CAS | 7333-63-3 |
| Molecular Formula | C23 H21 N3 O4 . H Cl |
| Quantity | 250 mg |
| Synonym | (4-Bromobutyl)triphenylphosphonium Bromide, (4-Bromobutyl)triphenyl-phosphonium Bromide, NSC 84071 |
| InChI Formula | InChI=1S/C23H21N3O4.ClH/c24-23(25)26-19-10-8-18(9-11-19)22(28)30-20-12-6-16(7-13-20)14-21(27)29-15-17-4-2-1-3-5-17;/h1-13H,14-15H2,(H4,24,25,26);1H |
| SMILES | Cl.NC(=N)Nc1ccc(cc1)C(=O)Oc2ccc(CC(=O)OCc3ccccc3)cc2 |
| IUPAC Name | [4-(2-oxo-2-phenylmethoxyethyl)phenyl] 4-carbamimidamidobenzoate;hydrochloride |
| Molecular Weight (g/mol) | 439.892 |
| Formula Weight | 439.13 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction