missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyl acetate, 97%

Product Code. 11447527
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447527 10 g 10g
11457527 50 g 50g
11467527 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11447527 Supplier Thermo Scientific Alfa Aesar Supplier No. A15632.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4753-59-7 | C6H11BrO2 | 195.056 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4753-59-7
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.056
MDL Number MFCD00000263
InChI Key UOABIRUEGSGTSA-UHFFFAOYSA-N
Synonym 4-bromobutylacetate, 4-bromo-n-butyl acetate, 1-butanol, 4-bromo-, acetate, acetic acid 4-bromobutyl ester, 1-butanol, 4-bromo-, 1-acetate, 4-bromo-1-acetoxybutane, 1-butanol, 4-bromo-,1-acetate, bromobutyl acetate, 4-acetoxy-butylbromide, 4-acetoxybutyl bromide
PubChem CID 78491
IUPAC Name 4-bromobutyl acetate
SMILES CC(=O)OCCCCBr

Specifications

Density 1.348
Boiling Point 96°C to 97°C (15 mmHg)
Flash Point 109°C (228°F)
Refractive Index 1.46
Quantity 10 g
Beilstein 1749695
Formula Weight 195.07
Percent Purity ≥98%
Chemical Name or Material 4-Bromobutyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.