missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromobutoxy)benzene, 97%

Product Code. p-7022266
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427157 5 g 5g
11437157 25 g 25g
11447157 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427157 Supplier Thermo Scientific Alfa Aesar Supplier No. A14956.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1200-03-9 | C10H13BrO | 229.12 g/mol

(4-Bromobutoxy)benzene is used to produce 5-phenoxy-valeric acid with carbon dioxide and reagent Mg.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(4-Bromobutoxy)benzene is used to produce 5-phenoxy-valeric acid with carbon dioxide and reagent Mg.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1200-03-9
Molecular Formula C10H13BrO
Molecular Weight (g/mol) 229.12
MDL Number MFCD00000262
InChI Key QBLISOIWPZSVIK-UHFFFAOYSA-N
Synonym 4-bromobutoxy benzene, 4-phenoxybutyl bromide, 4-bromobutyl phenyl ether, benzene, 4-bromobutoxy, 1-bromo-4-phenoxybutane, phenoxybutyl bromide, 4-phenoxybutylbromide, benzene, bromobutoxy, 4-bromo-1-phenoxybutane, 4-brombutoxy benzol
PubChem CID 70986
IUPAC Name 4-bromobutoxybenzene
SMILES BrCCCCOC1=CC=CC=C1

Specifications

Melting Point 40°C to 43°C
Boiling Point 153°C to 154°C (18 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Quantity 5 g
Beilstein 2044374
Solubility Information Insoluble in water.
Formula Weight 229.12
Percent Purity 97%
Chemical Name or Material (4-Bromobutoxy)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.