missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobiphenyl, TRC

Product Code. 30336998
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336998 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336998 Supplier Toronto Research Chemicals Supplier No. TRCB6805451G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material PBB-No. 3
CAS 92-66-0
Molecular Formula C12 H9 Br
Quantity 1 g
Synonym PBB 3 (4-Bromobiphenyl), 1,1'-Biphenyl, 4-bromo-, Biphenyl, 4-bromo- (6CI,8CI), 4-Bromo-1,1'-biphenyl, (4-Bromophenyl)benzene, 1-Bromo-4-phenylbenzene, 4-Biphenyl bromide, 4-Biphenylyl bromide, 4-Bromobiphenyl, 4-Bromodiphenyl, 4-Phenylbromobenzene, 4'-Bromobiphenyl, NSC 406933, PBB 3, p-Biphenylyl bromide, p-Bromobiphenyl, p-Bromodiphenyl, p-Bromophenylbenzene, p-Phenylbromobenzene, p-Phenylphenyl bromide
InChI Formula InChI=1S/C12H9Br/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H
SMILES Brc1ccc(cc1)c2ccccc2
IUPAC Name 1-bromo-4-phenylbenzene
Molecular Weight (g/mol) 233.1
Formula Weight 231.9888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.