missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15435395
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435395 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15435395 Supplier Thermo Scientific Alfa Aesar Supplier No. H58662.AD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 115055-85-1 | C7H6Br2Zn | 315.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 115055-85-1
Density 1.015
Molecular Formula C7H6Br2Zn
MDL Number MFCD01311435
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym zinc 2+ ion 4-bromophenyl methanide bromide
InChI Key HJQDBIKHGQSYJU-UHFFFAOYSA-M
SMILES [Zn++].[Br-].[CH2-]C1=CC=C(Br)C=C1
IUPAC Name zinc(2+) (4-bromophenyl)methanide bromide
Molecular Weight (g/mol) 315.31
PubChem CID 57346859
Formula Weight 315.31
Chemical Name or Material 4-Bromobenzylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.