missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromobenzyl)triphenylphosphonium bromide, 98%

Product Code. 11442248
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442248 5 g 5g
11452248 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11442248 Supplier Thermo Scientific Alfa Aesar Supplier No. A15220.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51044-13-4 | C25H21Br2P | 512.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51044-13-4
Molecular Formula C25H21Br2P
Molecular Weight (g/mol) 512.23
MDL Number MFCD00051922
InChI Key FQJYKXVQABPCRA-UHFFFAOYSA-M
Synonym 4-bromobenzyl triphenylphosphonium bromide, phosphonium, 4-bromophenyl methyl triphenyl-, bromide, 4-bromophenyl methyl triphenylphosphanium bromide, acmc-20apnl, 4-bromobenzyltriphenylphosphoniumbromide, 4-bromobenzyl triphenylphosphoniumbromide, 4-bromobenzyl triphenylphosphonium-bromide, 4-bromobenzyl triphenyl-phosphonium bromide, 4-bromobenzyl triphenyl phosphonium bromide
PubChem CID 2724858
IUPAC Name (4-bromophenyl)methyl-triphenylphosphanium;bromide
SMILES [Br-].BrC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

Melting Point 270°C to 273°C
Quantity 5 g
Beilstein 4173386
Sensitivity Hygroscopic
Formula Weight 512.24
Percent Purity 98%
Chemical Name or Material (4-Bromobenzyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.