missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl Cyanide, TRC

Product Code. 30342295
Change view
Click to view available options
Quantity:
25 g
250 g
Unit Size:
250g
25g
Product Code. Quantity unitSize
30342295 25 g 25g
30336182 250 g 250g
2 options
This item is not returnable. View return policy

Product Code. 30342295

Brand: Toronto Research Chemicals TRCB68021025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromobenzyl Cyanide
CAS 16532-79-9
Molecular Formula C8 H6 Br N
Quantity 25 g
Synonym (4-Bromophenyl)acetonitrile, (p-Bromophenyl)acetonitrile, 1-Bromo-4-cyanomethylbenzene, 2-(4-Bromophenyl)acetonitrile, 4-(Cyanomethyl)bromobenzene, 4-Bromobenzeneacetonitrile, 4-Bromobenzylnitrile, p-Bromobenzyl Cyanide, NSC 84174
InChI Formula InChI=1S/C8H6BrN/c9-8-3-1-7(2-4-8)5-6-10/h1-4H,5H2
SMILES Brc1ccc(CC#N)cc1
IUPAC Name 2-(4-bromophenyl)acetonitrile
Molecular Weight (g/mol) 196.044
Formula Weight 194.9684
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.