missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl chloride, 98%

Product Code. 10234991 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10234991 5 g Glass bottle 5g
10797192 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10234991 Supplier Thermo Scientific Acros Supplier No. 346750050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 589-17-3 | C7H6BrCl | 205.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-17-3
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00040867
InChI Key BSIIGUGKOPPTPZ-UHFFFAOYSA-N
Synonym 1-bromo-4-chloromethyl benzene, 4-bromobenzyl chloride, p-bromobenzyl chloride, 4-bromobenzylchloride, 4-bromo-alpha-chlorotoluene, benzene, 1-bromo-4-chloromethyl, ccris 5105, p-bromo-alpha-chlorotoluene, pubchem2478, 4-bromo-benzylchloride
PubChem CID 68528
IUPAC Name 1-bromo-4-(chloromethyl)benzene
SMILES ClCC1=CC=C(Br)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 36°C to 40°C
Color White
Boiling Point 136°C to 139°C (27.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2C6H4Br
Quantity 5 g
Merck Index 15, 1417
Solubility Information Solubility in water: insoluble in water
Formula Weight 205.48
Percent Purity 98%
Physical Form Powder or Granules
Chemical Name or Material 4-Bromobenzyl chloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Bromobenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.