missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl chloride, 97%

Product Code. 11362447
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362447 5 g 5g
11372447 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362447 Supplier Thermo Scientific Alfa Aesar Supplier No. L11486.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 589-17-3 | C7H6BrCl | 205.48 g/mol

4-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-17-3
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.48
MDL Number MFCD00040867
InChI Key BSIIGUGKOPPTPZ-UHFFFAOYSA-N
Synonym 1-bromo-4-chloromethyl benzene, 4-bromobenzyl chloride, p-bromobenzyl chloride, 4-bromobenzylchloride, 4-bromo-alpha-chlorotoluene, benzene, 1-bromo-4-chloromethyl, ccris 5105, p-bromo-alpha-chlorotoluene, pubchem2478, 4-bromo-benzylchloride
PubChem CID 68528
IUPAC Name 1-bromo-4-(chloromethyl)benzene
SMILES ClCC1=CC=C(Br)C=C1

Specifications

Melting Point 38°C to 41°C
Boiling Point 106°C to 107°C (10 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 6123220
Merck Index 14,1410
Solubility Information Insoluble in water.
Formula Weight 205.49
Percent Purity 97%
Chemical Name or Material 4-Bromobenzyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.