missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzyl Bromide, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | 4-Bromobenzyl Bromide |
| CAS | 589-15-1 |
| Molecular Formula | C22 H28 O4 |
| Quantity | 50 g |
| Synonym | p,alpha-Dibromo-toluene, 1-(Bromomethyl)-4-bromobenzene, 1-Bromo-4-(bromomethyl)benzene, 4-(Bromomethyl)bromobenzene, 4-Bromo-1-bromomethylbenzene, 4-Bromobenzyl bromide, NSC 113584, p-Bromobenzyl bromide, alpha,p-Dibromotoluene |
| InChI Formula | InChI=1S/C22H28O4/c1-4-22(26-13(2)23)10-8-19-17-12-20(25)18-11-14(24)5-6-15(18)16(17)7-9-21(19,22)3/h1,11,15-17,19-20,25H,5-10,12H2,2-3H3/t15-,16+,17-,19-,20-,21+,22?/m1/s1 |
| SMILES | CC(=O)OC1(CC[C@@H]2[C@@H]3C[C@@H](O)C4=CC(=O)CC[C@@H]4[C@@H]3CC[C@]12C)C#C |
| IUPAC Name | [(6R,8R,9R,10R,13S,14R)-17-ethynyl-6-hydroxy-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate |
| Molecular Weight (g/mol) | 356.455 |
| Formula Weight | 356.199 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction