missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl bromide, 98+%

Product Code. 11414253
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414253 25 g 25g
11424253 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414253 Supplier Thermo Scientific Alfa Aesar Supplier No. A10892.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 589-15-1 | C7H6Br2 | 249.933 g/mol

4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.

Solubility
Easily soluble in hot alcohol, ether, carbon disulfide, benzene and glacial acetic acid. Soluble in water and alcohol.

Notes
Keep containers tightly closed in a dry, cool and well-ventilated place. Store at 4°C. Incompatible with strong oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-15-1
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000179
InChI Key YLRBJYMANQKEAW-UHFFFAOYSA-N
Synonym 4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide
PubChem CID 68527
IUPAC Name 1-bromo-4-(bromomethyl)benzene
SMILES C1=CC(=CC=C1CBr)Br

Specifications

Melting Point 60°C to 64°C
Boiling Point 115°C to 124°C (12 mmHg)
Quantity 25 g
UN Number UN1759
Beilstein 606498
Merck Index 14,1409
Solubility Information Easily soluble in hot alcohol,ether,carbon disulfide,benzene and glacial acetic acid. Soluble in water and alcohol.
Formula Weight 249.94
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.