missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl bromide, 98%

Product Code. 10275340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10275340 25 g Glass bottle 25g
10563121 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10275340 Supplier Thermo Scientific Acros Supplier No. 150410250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 589-15-1 | C7H6Br2 | 249.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 589-15-1
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.93
MDL Number MFCD00000179
InChI Key YLRBJYMANQKEAW-UHFFFAOYSA-N
Synonym 4-bromobenzyl bromide, 1-bromo-4-bromomethyl benzene, p-bromobenzyl bromide, alpha,p-dibromotoluene, alpha,4-dibromotoluene, benzene, 1-bromo-4-bromomethyl, 4-bromobenzylbromide, unii-3d1ttl8bgn, a,p-dibromotoluene, 4-bromo benzyl bromide
PubChem CID 68527
IUPAC Name 1-bromo-4-(bromomethyl)benzene
SMILES C1=CC(=CC=C1CBr)Br

Specifikationer

CAS Min % 97.5
CAS Max % 100.0
Melting Point 60.0°C to 64.0°C
Color Tan to White
Boiling Point 115.0°C to 124.0°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH2Br
Quantity 25 g
Beilstein 05, 308
Merck Index 15, 1416
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in glacial acetic acid,soluble in alcohol,ether,carbon disulfide and,benzene
Formula Weight 249.93
Percent Purity 98%
Physical Form Coarse Crystalline Solid or Solid Melt
Chemical Name or Material 4-Bromobenzyl bromide, 98%
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 4-Bromobenzyl bromide
Signalord
  • Danger
Riskkategorier
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Faroangivelser
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.