missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzyl alcohol, 99%, Thermo Scientific Chemicals

Product Code. 10731461
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-75-6
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00004650
InChI Key VEDDBHYQWFOITD-UHFFFAOYSA-N
Synonym 4-bromobenzyl alcohol, 4-bromophenyl methanol, p-bromobenzyl alcohol, benzenemethanol, 4-bromo, para-bromobenzyl alcohol, 4-bromobenzylalcohol, p-bromo-benzyl alcohol, ccris 5119, 4-bromophenyl-methanol, benzyl alcohol, p-bromo
PubChem CID 70119
IUPAC Name (4-bromophenyl)methanol
SMILES C1=CC(=CC=C1CO)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 75.0°C to 78.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H4CH2OH
Quantity 10 g
Beilstein 06, 446
Formula Weight 187.04
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromobenzyl alcohol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'