missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzotrifluoride, 99%

Product Code. 10043953 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10043953 100 mL 100mL
10761631 5 mL 5mL
10761821 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10043953 Supplier Thermo Scientific Acros Supplier No. 166671000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 402-43-7 | C7H4BrF3 | 225.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-43-7
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.01
MDL Number MFCD00000398
InChI Key XLQSXGGDTHANLN-UHFFFAOYSA-N
Synonym 4-bromobenzotrifluoride, 1-bromo-4-trifluoromethyl benzene, benzene, 1-bromo-4-trifluoromethyl, p-bromobenzotrifluoride, 4-bromotrifluorotoluene, 4-bromo-alpha,alpha,alpha-trifluorotoluene, p-bromotrifluoromethylbenzene, 4-trifluoromethyl bromobenzene, 4-bromo-a,a,a-trifluorotoluene, 4-trifluoromethylbromobenzene
PubChem CID 67872
IUPAC Name 1-bromo-4-(trifluoromethyl)benzene
SMILES C1=CC(=CC=C1C(F)(F)F)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6000g/mL
Boiling Point 154.0°C to 155.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula BrC6H4CF3
Refractive Index 1.4720 to 1.4740
Quantity 100 mL
Specific Gravity 1.6
Formula Weight 225.01
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Bromobenzotrifluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Bromobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.