missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzenesulfonyl chloride, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Especificaciones
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | 74°C to 77°C |
| Color | White to Beige |
| Boiling Point | 153°C (15.0 mmHg) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (Ex CL) (Argentometry) |
| Packaging | Glass bottle |
| Linear Formula | BrC6H4SO2Cl |
| Quantity | 25 g |
| Mostrar más |
Seguridad y manipulación
Nombre del producto
- 4-Bromobenzenesulfonyl chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido