missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzenesulfinic acid sodium salt dihydrate, 97%

Product Code. 11429786
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429786 2.5 g 2.5g
11439786 10 g 10g
11449786 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429786 Supplier Thermo Scientific Alfa Aesar Supplier No. B20281.88
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 175278-64-5 | C6H4BrO2S | 220.06 g/mol

4-Bromobenzenesulfinic acid sodium salt dehydrate is used in the preparation of unsymmetrical internal alkynes and vinyl sulfones by reaction with alkynes by using palladium as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzenesulfinic acid sodium salt dehydrate is used in the preparation of unsymmetrical internal alkynes and vinyl sulfones by reaction with alkynes by using palladium as a catalyst.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175278-64-5
Molecular Formula C6H4BrO2S
Molecular Weight (g/mol) 220.06
MDL Number MFCD00210139
InChI Key QBLQHDHWTVMHRH-UHFFFAOYSA-M
Synonym sodium 4-bromobenzenesulfinate dihydrate, 4-bromobenzenesulfinic acid sodium salt dihydrate, sodium 4-bromobenzene-1-sulphinate dihydrate, acmc-209e9v, c6h4bro2s.na.2h2o, sodium 4-bromophenylsulfinate dihydrate, sodium 4-bromobenzenesulfinate, dihydrate, sodium 4-bromanylbenzenesulfinate dihydrate, 4-bromobenzenesulfonic acid sodium salt 2-hydrate, sodium 4-bromobenzene-1-sulfinate-water 1/1/2
PubChem CID 23683129
IUPAC Name sodium;4-bromobenzenesulfinate;dihydrate
SMILES [O-]S(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point >280°C
Quantity 2.5 g
Solubility Information Soluble in water.
Formula Weight 279.09 (243.06 Anhydrous)
Percent Purity 97%
Chemical Name or Material 4-Bromobenzenesulfinic acid sodium salt dihydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.