missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzenediazonium tetrafluoroborate, 96%

Product Code. 11491687
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11491687 5 g 5g
11401697 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11491687

Brand: Thermo Scientific Alfa Aesar B25670.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 673-40-5 | C6H4BBrF4N2 | 270.82 g/mol

4-Bromobenzenediazonium tetrafluoroborate was used in preparation of surface modified Au(111) electrode, 1-(4-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzenediazonium tetrafluoroborate was used in preparation of surface modified Au(111) electrode, 1-(4′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 673-40-5
Molecular Formula C6H4BBrF4N2
Molecular Weight (g/mol) 270.82
MDL Number MFCD00011894
InChI Key FXTCQUCYDJZGGU-UHFFFAOYSA-N
Synonym 4-bromobenzenediazonium tetrafluoroborate, acmc-209nyn, 4-bromobenzene-1-diazonium; tetrafluoroboranuide, boron +3 cation; 4-bromobenzenediazonium; tetrafluoride
PubChem CID 2734810
IUPAC Name 4-bromobenzenediazonium;tetrafluoroborate
SMILES F[B-](F)(F)F.BrC1=CC=C(C=C1)[N+]#N

Specifications

Melting Point ∼135°C (decomposition)
Quantity 5 g
UN Number UN1759
Sensitivity Light and moisture sensitive
Formula Weight 270.83
Percent Purity 96%
Chemical Name or Material 4-Bromobenzenediazonium tetrafluoroborate
compliance-icons compliance-icons
Product Identifier
  • 4-Bromobenzenediazonium tetrafluoroborate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-Bromobenzenediazonium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.