missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzamide, 97%

Product Code. 11475777
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475777 50 g 50g
11455777 2 g 2g
11465777 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475777 Supplier Thermo Scientific Alfa Aesar Supplier No. B24348.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 698-67-9 | C7H6BrNO | 200.035 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-67-9
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD00007991
InChI Key ZRWNRAJCPNLYAK-UHFFFAOYSA-N
Synonym p-bromobenzamide, benzamide, 4-bromo, benzamide, p-bromo, p-bromobenzoic acid amide, 4-bromo-benzamide, 4-bromo benzamide, pubchem3737, acmc-209obe, 4-bromobenzamide, 4-bromobenzoic acid amide
PubChem CID 69683
IUPAC Name 4-bromobenzamide
SMILES C1=CC(=CC=C1C(=O)N)Br

Specifications

Melting Point 190°C to 193°C
Quantity 50 g
Formula Weight 200.03
Percent Purity 97%
Chemical Name or Material 4-Bromobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.