missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzamide, 97%, Thermo Scientific™

Product Code. 10713001 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10713001 10 g Glass bottle 10g
10104160 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10713001 Supplier Thermo Scientific Acros Supplier No. 184140100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 698-67-9
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.03
MDL Number MFCD00007991
InChI Key ZRWNRAJCPNLYAK-UHFFFAOYSA-N
Synonym p-bromobenzamide, benzamide, 4-bromo, benzamide, p-bromo, p-bromobenzoic acid amide, 4-bromo-benzamide, 4-bromo benzamide, pubchem3737, acmc-209obe, 4-bromobenzamide, 4-bromobenzoic acid amide
PubChem CID 69683
IUPAC Name 4-bromobenzamide
SMILES C1=CC(=CC=C1C(=O)N)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 189°C to 194°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula BrC6H4CONH2
Quantity 10 g
Beilstein 09, 353
Formula Weight 200.03
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Bromobenzamide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromobenzamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.