missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzaldehyde, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 4-Bromobenzaldehyde |
| CAS | 1122-91-4 |
| Molecular Formula | C7H5BrO |
| Quantity | 10 g |
| Synonym | 1-Bromo-4-formylbenzene, 4-Formyl-1-bromobenzene, 4-Formylbromobenzene, 4-Formylphenyl Bromide, NSC 21638, p-Bromobenzaldehyde, p-Formylbromobenzene |
| InChI Formula | InChI=1S/C7H5BrO/c8-7-3-1-6(5-9)2-4-7/h1-5H |
| SMILES | Brc1ccc(C=O)cc1 |
| IUPAC Name | 4-bromobenzaldehyde |
| Molecular Weight (g/mol) | 185.02 |
| Formula Weight | 183.9524 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction