missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzaldehyde diethyl acetal, 99%, Thermo Scientific™

Product Code. 10296373 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10296373 50 g Glass bottle 50g
10553624 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10296373 Supplier Thermo Scientific Acros Supplier No. 390020500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 34421-94-8
Molecular Formula C11H15BrO2
Molecular Weight (g/mol) 259.14
MDL Number MFCD01863514
InChI Key BFSNEBVTOODGHZ-UHFFFAOYSA-N
Synonym 4-bromobenzaldehyde diethyl acetal, 1-bromo-4-diethoxymethyl benzene, 4-bromobenzaldehydediethylacetal, benzene, 1-bromo-4-diethoxymethyl, 4-bromophenyl diethoxymethane, 4-bromobenzaldehyde diethylacetal, 4-bromo-alpha,alpha-diethoxytoluene, pubchem5372, acmc-1aigt, 4-bromobenzaldehydediethyl acetal
PubChem CID 688340
IUPAC Name 1-bromo-4-(diethoxymethyl)benzene
SMILES CCOC(OCC)C1=CC=C(Br)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.274g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H4CH(OCH2CH3)2
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
Quantity 50 g
Formula Weight 259.14
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 4-Bromobenzaldehyde diethyl acetal
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromobenzaldehyde diethyl acetal
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.