missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzaldehyde, 99%

Product Code. 11380270 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11380270

Brand: Thermo Scientific Acros 106675000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1122-91-4 | C7H5BrO | 185.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-91-4
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
InChI Key ZRYZBQLXDKPBDU-UHFFFAOYSA-N
Synonym p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd
PubChem CID 70741
IUPAC Name 4-bromobenzaldehyde
SMILES C1=CC(=CC=C1C=O)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 55.0°C to 58.0°C
Color White
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CHO
Quantity 500 g
Beilstein 07,239
Solubility Information Solubility in water: insoluble
Formula Weight 185.02
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 4-Bromobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.