missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzaldehyde, 98+%

Product Code. 11412357
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412357 25 g 25g
11422357 100 g 100g
11432357 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412357 Supplier Thermo Scientific Alfa Aesar Supplier No. A14237.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1122-91-4 | C7H5BrO | 185.02 g/mol

4-Bromobenzaldehyde is widely utilized as an intermediate for the preparation of agrochemicals, pharmaceuticals and other organic compounds. It is used as a precursor and reacts with sulfamethoxazole to prepare Schiff’s base, 4-[(4-Bromo-benzylidene)-amino]-N-(5-methyl-isoxazol-3-yl)-benzene sulfonamide, which can be used for the photo stability of polyvinyl chloride (PVC). It plays an important role in palladium-catalyzed mono- and bis-arylation of 3,4-(ethylenedioxy)thiophenes. It is employed in a cross-coupling study with potassium vinyltrifluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzaldehyde is widely utilized as an intermediate for the preparation of agrochemicals, pharmaceuticals and other organic compounds. It is used as a precursor and reacts with sulfamethoxazole to prepare Schiff’s base, 4-[(4-Bromo-benzylidene)-amino]-N-(5-methyl-isoxazol-3-yl)-benzene sulfonamide, which can be used for the photo stability of polyvinyl chloride (PVC). It plays an important role in palladium-catalyzed mono- and bis-arylation of 3,4-(ethylenedioxy)thiophenes. It is employed in a cross-coupling study with potassium vinyltrifluoroborate.

Solubility
Soluble in ethanol and benzene. Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-91-4
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00003377
InChI Key ZRYZBQLXDKPBDU-UHFFFAOYSA-N
Synonym p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd
PubChem CID 70741
IUPAC Name 4-bromobenzaldehyde
SMILES C1=CC(=CC=C1C=O)Br

Specifications

Melting Point 55°C to 58°C
Boiling Point 66°C to 68°C (2 mmHg)
Flash Point 108°C (226°F)
Odor Characteristic
Quantity 25 g
Beilstein 507100
Sensitivity Air Sensitive
Solubility Information Soluble in ethanol and benzene. Insoluble in water.
Formula Weight 185.03
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.