missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoanisole(p-Methoxyphenyl Bromide), TRC

Product Code. 30464809
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464809 5 g 5g
30467109 25 g 25g
30480767 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30464809 Supplier Toronto Research Chemicals Supplier No. TRCM2658155G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-92-7
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.03
Synonym 1-Bromo-4-methoxybenzene, 1-Methoxy-4-bromobenzene, 4-Anisyl bromide, 4-Bromo-1-methoxybenzene, 4-Bromoanisol, 4-Bromomethoxybenzene, 4-Methoxy-1-bromobenzene, 4-Methoxybromobenzene, 4-Methoxyphenyl bromide, NSC 8042, p-Anisyl bromide, p-Bromanisole, p-Bromoanisole, p-Bromomethoxybenzene, p-Bromophenyl methyl ether, p-Methoxybromobenzene
IUPAC Name 1-bromo-4-methoxybenzene
SMILES COc1ccc(Br)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H7BrO/c1-9-7-4-2-6(8)3-5-7/h2-5H,1H3
Formula Weight 185.968
Chemical Name or Material 4-Bromoanisole(p-Methoxyphenyl Bromide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.