missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoaniline, 98+%

Product Code. p-7022379
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11482088

Brand: Thermo Scientific Alfa Aesar A15126.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-40-1 | C6H6BrN | 172.025 g/mol

4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.

Solubility
Soluble in alcohol and ether. Insoluble in cold water.

Notes
Store in a cool place. Light sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-40-1
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.025
MDL Number MFCD00007822
InChI Key WDFQBORIUYODSI-UHFFFAOYSA-N
Synonym p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine
PubChem CID 7807
IUPAC Name 4-bromoaniline
SMILES C1=CC(=CC=C1N)Br

Specifications

Melting Point 60°C to 65°C
Boiling Point 222°C to 224°C
Assay Percent Range ≥98%
Quantity 50 g
UN Number UN2811
Beilstein 742031
Merck Index 14,1404
Solubility Information Soluble in alcohol and ether. Insoluble in cold water.
Formula Weight 172.03
Percent Purity ≥98%
Chemical Name or Material 4-Bromoaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Bromoaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Bromoaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.