missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromoacetyl)pyridine hydrobromide, 98%

Product Code. 15434997
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15434997 1 g 1g
15444997 5 g 5g
15454997 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15434997 Supplier Thermo Scientific Alfa Aesar Supplier No. H32285.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5349-17-7 | C7H7Br2NO | 280.947 g/mol

Design and synthesis of potent, selective, and orally bioavailable tetrasubstituted imidazole inhibitors of p38 mitogen-activated protein kinase 4-(Bromoacetyl)pyridine hydrobromide was used to prepare 2-Phenyl-4-(4-pyridyl)imidazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Design and synthesis of potent, selective, and orally bioavailable tetrasubstituted imidazole inhibitors of p38 mitogen-activated protein kinase 4-(Bromoacetyl)pyridine hydrobromide was used to prepare 2-Phenyl-4-(4-pyridyl)imidazole.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5349-17-7
Molecular Formula C7H7Br2NO
Molecular Weight (g/mol) 280.947
MDL Number MFCD02681893
InChI Key RGALBQILADNMKA-UHFFFAOYSA-N
Synonym 4-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-4-yl ethanone hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hydrobromide, 2-bromo-1-pyridin-4-yl ethan-1-one hydrobromide, ethanone, 2-bromo-1-4-pyridinyl-, hydrobromide, 4-2-bromoacetyl pyridine hydrobromide, 4-bromoacetylpyridine hydrobromide, 2-bromo-1-pyrdin-4-ylethan-1-one hydrobromide, 2-bromo-1-4-pyridinyl-1-ethanone hbr, 4-bromoacetyl pyridine hbr
PubChem CID 2776239
IUPAC Name 2-bromo-1-pyridin-4-ylethanone;hydrobromide
SMILES C1=CN=CC=C1C(=O)CBr.Br

Specifications

Melting Point ∽200°C (decomposition)
Odor Weak
Quantity 1 g
UN Number UN3261
Solubility Information Soluble in water.
Formula Weight 280.95
Percent Purity 98%
Chemical Name or Material 4-(Bromoacetyl)pyridine hydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.