missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromoacetanilide, 98%

Product Code. 11397895
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397895 250 g 250g
11387895 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397895 Supplier Thermo Scientific Alfa Aesar Supplier No. L02924.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-88-8 | C8H8BrNO | 214.06 g/mol

4'-Bromoacetanilide is used as an internal standard for phenylurea and the respective metabolic products in oysters. It has also been used as a reagent in the production of a new ligand to form a coordination compex between gadolinium(III) and T=titanium(IV) oxide. It was used in the synthesis of new ligand for anchoring Gd(III) chelates onto TiO2 surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Bromoacetanilide is used as an internal standard for phenylurea and the respective metabolic products in oysters. It has also been used as a reagent in the production of a new ligand to form a coordination compex between gadolinium(III) and T=titanium(IV) oxide. It was used in the synthesis of new ligand for anchoring Gd(III) chelates onto TiO2 surface.

Solubility
Moderately soluble in alcohol. Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-88-8
Molecular Formula C8H8BrNO
Molecular Weight (g/mol) 214.06
MDL Number MFCD00000092
InChI Key MSLICLMCQYQNPK-UHFFFAOYSA-N
Synonym 4'-bromoacetanilide, n-4-bromophenyl acetamide, p-bromoacetanilide, 4-bromoacetanilide, bromoanilide, acetamide, n-4-bromophenyl, antisepsin, bromoantifebrin, asepsin, p-bromo-n-acetanilide
PubChem CID 7683
IUPAC Name N-(4-bromophenyl)acetamide
SMILES CC(=O)NC1=CC=C(Br)C=C1

Specifications

Melting Point 166°C to 171°C
Odor Odorless
Quantity 250 g
Beilstein 2208091
Merck Index 14,1399
Solubility Information Moderately soluble in alcohol. Insoluble in water.
Formula Weight 214.06
Percent Purity 98%
Chemical Name or Material 4'-Bromoacetanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.