missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-o-xylene, 97%

Product Code. p-7022117
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11487776

Brand: Thermo Scientific Alfa Aesar A14736.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 583-71-1 | C8H9Br | 185.064 g/mol

4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-71-1
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000086
InChI Key QOGHRLGTXVMRLM-UHFFFAOYSA-N
Synonym 4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo
PubChem CID 68504
IUPAC Name 4-bromo-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)Br)C

Specifications

Density 1.37
Boiling Point 214°C to 215°C
Quantity 5 g
Refractive Index 1.556
Beilstein 2040354
Solubility Information Not miscible in water.
Formula Weight 185.07
Percent Purity 97%
Chemical Name or Material 4-Bromo-o-xylene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.