missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N,N-dimethylaniline, 99%

Product Code. 10256370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10256370 100 g Plastic bottle 100g
10449920 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10256370 Supplier Thermo Scientific Acros Supplier No. 222211000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 586-77-6 | C8H10BrN | 200.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-77-6
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00000093
InChI Key XYZWMVYYUIMRIZ-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-bromoaniline, 4-dimethylaminobromobenzene, p-bromo-n,n-dimethylaniline, benzenamine, 4-bromo-n,n-dimethyl, n,n-dimethyl-4-bromoaniline, p-dimethylamino phenyl bromide, p-dimethylaminobromobenzene, 1-bromo-4-dimethylamino benzene, p-bromo dimethylamino benzene, p-n,n-dimethylaminobromobenzene
PubChem CID 11465
IUPAC Name 4-bromo-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(Br)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53°C to 56°C
Color Blue or Gray to White
Boiling Point 264°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula BrC6H4N(CH3)2
Quantity 100 g
Beilstein 12, 637
Solubility Information Solubility in water: insoluble. Other solubilities: methanol 100g/L
Formula Weight 200.08
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 4-Bromo-N, N-dimethylaniline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-N,N-dimethylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.