missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-(3-methylphenyl)benzamide, 97%, Thermo Scientific™

Catalog Number 15423898
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
81636-17-1
C14H12BrNO
290.16
MRYWJNCVXBZMKZ-UHFFFAOYSA-N
4-bromo-n-3-methylphenyl benzamide, benzamide, n-3-methylphenyl-4-bromo, 4-bromo-n-m-tolyl-benzamide, 4-bromo-n-m-tolyl benzamide, 4-bromo-n∼1∼-3-methylphenyl benzamide, 4-bromophenyl-n-3-methylphenyl carboxamide
532151
4-bromo-N-(3-methylphenyl)benzamide
CC1=CC(=CC=C1)NC(=O)C2=CC=C(C=C2)Br
This item is not returnable. View return policy
81636-17-1
C14H12BrNO
290.16
MRYWJNCVXBZMKZ-UHFFFAOYSA-N
4-bromo-n-3-methylphenyl benzamide, benzamide, n-3-methylphenyl-4-bromo, 4-bromo-n-m-tolyl-benzamide, 4-bromo-n-m-tolyl benzamide, 4-bromo-n∼1∼-3-methylphenyl benzamide, 4-bromophenyl-n-3-methylphenyl carboxamide
532151
4-bromo-N-(3-methylphenyl)benzamide
CC1=CC(=CC=C1)NC(=O)C2=CC=C(C=C2)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 290.15
Percent Purity 97%
Chemical Name or Material 4-Bromo-N-(3-methylphenyl)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'