missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-DL-phenylglycine, 95%, Thermo Scientific Chemicals

Product Code. 15476747
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476747 1 g 1g
15486747 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476747 Supplier Thermo Scientific Alfa Aesar Supplier No. H33865.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119397-06-7
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.06
MDL Number MFCD00049327
InChI Key APLQICUORRMFHY-UHFFFAOYNA-N
Synonym 2-amino-2-4-bromophenyl acetic acid, amino 4-bromophenyl acetic acid, amino-4-bromo-phenyl-acetic acid, 2-4-bromophenyl glycine, 4-bromo-dl-phenylglycine, 2-amino-2-4-bromophenyl aceticacid, dl-4-br-phg-oh, 2-4-bromophenyl-dl-glycine, +-2-amino-2-4-bromophenyl acetic acid
PubChem CID 2764157
IUPAC Name 2-amino-2-(4-bromophenyl)acetic acid
SMILES NC(C(O)=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 273°C to 274°C
Quantity 1 g
Formula Weight 230.1
Percent Purity 95%
Chemical Name or Material 4-Bromo-DL-phenylglycine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.