missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-6-(trifluoromethyl)-1H-benzo[d]imidazole, 97%, Thermo Scientific™

Product Code. 10437602
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10437602

Brand: Thermo Scientific Maybridge BTB05258DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175135-14-5
Molecular Formula C8H4BrF3N2
Molecular Weight (g/mol) 265.033
MDL Number MFCD00067734
InChI Key HYTQERQCUFICAX-UHFFFAOYSA-N
Synonym 4-bromo-6-trifluoromethyl-1h-benzo d imidazole, 4-bromo-6-trifluoromethyl benzimidazole, 4-bromo-6-trifluoromethyl-1h-benzimidazole, 4-bromo-6-trifluoromethyl-1h-1,3-benzodiazole, 1h-benzimidazole,7-bromo-5-trifluoromethyl, 7-bromo-5-trifluoromethyl-1h-benzo d imidazole, 4-bromo-6-trifluoromethyl-3h-1,3-benzodiazole, pubchem6508, buttpark 95\04-47, 4-bromo-6-trifluoromethylbenzimidazole
PubChem CID 2736427
IUPAC Name 4-bromo-6-(trifluoromethyl)-1H-benzimidazole
SMILES C1=C(C=C(C2=C1NC=N2)Br)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 265.03
Percent Purity 97%
Chemical Name or Material 4-Bromo-6-(trifluoromethyl)-1H-benzo[d]imidazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.