missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-6-(trifluoromethyl)-1H-benzo[d]imidazole, 97%, Thermo Scientific™

Product Code. 10334222
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10334222 10 g Amber Glass Bottle 10g
11340527 250 mg Amber Glass Bottle 250mg
10437602 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10334222 Supplier Thermo Scientific Maybridge Supplier No. BTB05258EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175135-14-5
Molecular Formula C8H4BrF3N2
Molecular Weight (g/mol) 265.033
MDL Number MFCD00067734
InChI Key HYTQERQCUFICAX-UHFFFAOYSA-N
Synonym 4-bromo-6-trifluoromethyl-1h-benzo d imidazole, 4-bromo-6-trifluoromethyl benzimidazole, 4-bromo-6-trifluoromethyl-1h-benzimidazole, 4-bromo-6-trifluoromethyl-1h-1,3-benzodiazole, 1h-benzimidazole,7-bromo-5-trifluoromethyl, 7-bromo-5-trifluoromethyl-1h-benzo d imidazole, 4-bromo-6-trifluoromethyl-3h-1,3-benzodiazole, pubchem6508, buttpark 95\04-47, 4-bromo-6-trifluoromethylbenzimidazole
PubChem CID 2736427
IUPAC Name 4-bromo-6-(trifluoromethyl)-1H-benzimidazole
SMILES C1=C(C=C(C2=C1NC=N2)Br)C(F)(F)F

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 265.03
Percent Purity 97%
Chemical Name or Material 4-Bromo-6-(trifluoromethyl)-1H-benzo[d]imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.