missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-5-chlorothiophene-2-sulfonyl chloride, ≥95%, Thermo Scientific™

Product Code. 10307404
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10307404 10 g 10g
10640923 250 mg 250mg
10283082 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10307404 Supplier Thermo Scientific Maybridge Supplier No. SEW02652EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 166964-35-8
Molecular Formula C4HBrCl2O2S2
Molecular Weight (g/mol) 295.974
MDL Number MFCD00052094
InChI Key PABOKJJDABSQCK-UHFFFAOYSA-N
Synonym 3-bromo-2-chlorothiophene-5-sulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulphonyl chloride, 4-bromo-5-chloro-2-thiophenesulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulphonylchloride, 3-bromo-2-chlorothiophene-5-sulphonyl chloride, 2-thiophenesulfonylchloride, 4-bromo-5-chloro, pubchem5475, 4-bromo-5-chloro-thiophene-2-sulfonyl chloride, 4-bromo-5-chlorothiophene-2-sulfonyl, 4-bromanyl-5-chloranyl-thiophene-2-sulfonyl chloride
PubChem CID 2797220
IUPAC Name 4-bromo-5-chlorothiophene-2-sulfonyl chloride
SMILES C1=C(SC(=C1Br)Cl)S(=O)(=O)Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 295.99
Percent Purity 95%
Chemical Name or Material 4-Bromo-5-chlorothiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.