missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-3'-(trifluoromethyl)acetanilide, 97%

Product Code. 11418797
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
11418797 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 11418797

Brand: Thermo Scientific Alfa Aesar A19939.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 41513-05-7 | C9H7BrF3NO | 282.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41513-05-7
Molecular Formula C9H7BrF3NO
Molecular Weight (g/mol) 282.06
MDL Number MFCD00052361
InChI Key AOHSBSXIGFIMNC-UHFFFAOYSA-N
Synonym n-4-bromo-3-trifluoromethyl phenyl acetamide, 5-acetamido-2-bromobenzotrifluoride, 4-bromo-3-trifluoromethyl acetanilide, 4'-bromo-3'-trifluoromethyl acetanilide, n-4-bromo-3-trifluoromethyl phenyl acet amide, n1-4-bromo-3-trifluoromethyl phenyl acetamide, pubchem2757, maybridge4_003018, buttpark 37\11-74
PubChem CID 697838
IUPAC Name N-[4-bromo-3-(trifluoromethyl)phenyl]acetamide
SMILES CC(=O)NC1=CC(=C(C=C1)Br)C(F)(F)F

Specifications

Melting Point 116°C to 119°C
Quantity 10 g
Formula Weight 282.07
Percent Purity 97%
Chemical Name or Material 4'-Bromo-3'-(trifluoromethyl)acetanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.