missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-nitrotoluene, 98+%

Product Code. 11459572
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459572 25 g 25g
11469572 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459572 Supplier Thermo Scientific Alfa Aesar Supplier No. A10718.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5326-34-1 | C7H6BrNO2 | 216.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5326-34-1
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00024180
InChI Key UPBUTKQMDPHQAQ-UHFFFAOYSA-N
Synonym 4-bromo-3-nitrotoluene, 2-bromo-5-methylnitrobenzene, benzene, 1-bromo-4-methyl-2-nitro, bromonitrotoluene-4, 1-bromo-4-methyl-2-nitro-benzene, 3-nitro-4-bromotoluene, pubchem9585, acmc-209l5d, ksc490m7t, tpc-i121
PubChem CID 79224
IUPAC Name 1-bromo-4-methyl-2-nitrobenzene
SMILES CC1=CC=C(Br)C(=C1)[N+]([O-])=O

Specifications

Melting Point 29°C to 32°C
Density 1.578
Boiling Point 96°C (1 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 1945925
Sensitivity Light sensitive
Formula Weight 216.04
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-3-nitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.