missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylpyrazole, 97%, Thermo Scientific™

Product Code. 10663362 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10663362 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10663362 Supplier Thermo Scientific Acros Supplier No. 344350010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 13808-64-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 72°C to 79°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C4H5BrN2
MDL Number MFCD00005241
Quantity 1 g
Synonym 4-bromo-3-methylpyrazole, 4-bromo-3-methyl-1h-pyrazole, 3-methyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3-methyl, 4-bromo-3-methyl-2h-pyrazole, 4-bromo-3-methyl pyrazole, 3-methyl-4-bromo-1h-pyrazole, pubchem22533, timtec-bb sbb003957, acmc-1buv4
InChI Key IXQPRETWBGVNPJ-UHFFFAOYSA-N
SMILES CC1=C(C=NN1)Br
IUPAC Name 4-bromo-5-methyl-1H-pyrazole
Molecular Weight (g/mol) 161
PubChem CID 83741
Formula Weight 161
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Bromo-3-methylpyrazole
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-3-methylpyrazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.