missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylbenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 10813011
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-28-1
Molecular Formula C8H6BrO2
Molecular Weight (g/mol) 214.04
MDL Number MFCD00039526
InChI Key KWVXDZLVCISXIB-UHFFFAOYSA-M
Synonym 4-bromo-m-toluic acid, 4-bromo-3-methyl-benzoic acid, 4-bromo-3-methylbenzoicacid, 3-methyl-4-bromobenzoic acid, 4-bromo-3-methyl benzoic acid, benzoic acid, 4-bromo-3-methyl, pubchem3807, 4bromo-3-methylbenzoic acid, 4-bromo-5-methylbenzoic acid, ksc379g0b
PubChem CID 82131
IUPAC Name 4-bromo-3-methylbenzoic acid
SMILES CC1=CC(=CC=C1Br)C([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 215.5°C to 216.4°C
Color Tan to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 215.05
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Bromo-3-methylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-3-methylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'