missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylbenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. p-4323339 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11944591 1 g 1g
11954591 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11944591 Supplier Thermo Scientific Acros Supplier No. 442390010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 72256-93-0
Molecular Formula C7H6BrClO2S
Molecular Weight (g/mol) 269.54
MDL Number MFCD03094696
InChI Key JRNDXUKMWFVEIC-UHFFFAOYSA-N
Synonym 4-bromo-3-methylbenzene-1-sulfonyl chloride, 4-bromo-3-methylbenzenesulphonyl chloride, 4-bromo-3-methylbenzenesulphonylchloride, 4-bromo-3-methyl-benzenesulfonyl chloride, 6-bromo-m-toluene-3-sulfonyl chloride, pubchem15805, acmc-1bho4, jrndxukmwfveic-uhfffaoysa, 4-bromo-3-methylphenyl chlorosulfone, 4-bromo-3-methylbenzene sulfonyl chloride
PubChem CID 2737503
IUPAC Name 4-bromo-3-methylbenzenesulfonyl chloride
SMILES CC1=C(Br)C=CC(=C1)S(Cl)(=O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 58°C to 60°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 269.55
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-methylbenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-3-methylbenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.