missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylaniline, 97%, Thermo Scientific™

Product Code. 10032170 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10032170 10 g 10g
10696752 50 g 50g
15229666 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10032170 Supplier Thermo Scientific Acros Supplier No. 167500100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6933-10-4
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00007828

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 79.0°C to 83.0°C
Color Brown to White
Boiling Point 240.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(CH3)NH2
Quantity 10 g
Beilstein 12, 873
Synonym 4-bromo-m-toluidine, 5-amino-2-bromotoluene, 3-methyl-4-bromoaniline, benzenamine, 4-bromo-3-methyl, 4-bromo-3-methyl-phenylamine, 4-bromo-3-methyl-aniline, 4-bromo-3-methylbenzenamine, 4-bromo-3-methylphenylamine, pubchem4339, 4-bromo3-methylaniline
Solubility Information Solubility in water: insoluble
InChI Key MMEGELSFOYDPQW-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)N)Br
IUPAC Name 4-bromo-3-methylaniline
PubChem CID 23359
Formula Weight 186.05
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Bromo-3-methylaniline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-3-methylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.