missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylaniline, 97%, Thermo Scientific™

Product Code. 15229666 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15229666 250 g 250g
10032170 10 g 10g
10696752 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15229666 Supplier Thermo Scientific Acros Supplier No. 167502500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6933-10-4
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.05
MDL Number MFCD00007828

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 79.0°C to 83.0°C
Color Brown to White
Boiling Point 240.0°C
Assay Percent Range 96% min. (GC)
Linear Formula BrC6H3(CH3)NH2
Quantity 250 g
Beilstein 12, 873
Synonym methyl 2-aminoisonicotinate, 2-aminopyridine-4-carboxylic acid methyl ester, 2-aminoisonicotinic acid methyl ester, 4-methoxycarbonyl pyridin-2-amine, 4-pyridinecarboxylic acid, 2-amino-, methyl ester, methyl 2-amino-4-pyridinecarboxylate, methyl-2-aminopyridine-4-carboxylate, 2-amino-4-pyridinecarboxylic acid methylester, 2-amino-4-pyridinecarboxylic acid methyl ester, methyl-2-aminoisonicotinat
Solubility Information Solubility in water: insoluble
InChI Key SVWWNEYBEFASMP-UHFFFAOYSA-N
SMILES COC(=O)C1=CC(=NC=C1)N
IUPAC Name methyl 2-aminopyridine-4-carboxylate
PubChem CID 236984
Formula Weight 186.05
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Bromo-3-methylaniline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-3-methylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.