missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-3'-methylacetanilide, 98%, Thermo Scientific™

Catalog Number p-7026148
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
90914-81-1
C9H10BrNO
228.089
MFCD00040822
BYZHUFNLXFFINU-UHFFFAOYSA-N
n-4-bromo-3-methylphenyl acetamide, 4'-bromo-3'-methylacetanilide, 4-bromo-3-methylacetanilide, n-acetyl-4-bromo-m-toluidine, 3-methyl-4-bromoacetanilide, n-acetyl 4-bromo-3-methylaniline, n-4-bromo-3-methylphenyl-acetamide, maybridge1_005107, acmc-209r7e
738336
N-(4-bromo-3-methylphenyl)acetamide
CC1=C(C=CC(=C1)NC(=O)C)Br
This item is not returnable. View return policy
90914-81-1
C9H10BrNO
228.089
MFCD00040822
BYZHUFNLXFFINU-UHFFFAOYSA-N
n-4-bromo-3-methylphenyl acetamide, 4'-bromo-3'-methylacetanilide, 4-bromo-3-methylacetanilide, n-acetyl-4-bromo-m-toluidine, 3-methyl-4-bromoacetanilide, n-acetyl 4-bromo-3-methylaniline, n-4-bromo-3-methylphenyl-acetamide, maybridge1_005107, acmc-209r7e
738336
N-(4-bromo-3-methylphenyl)acetamide
CC1=C(C=CC(=C1)NC(=O)C)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 102°C to 104°C
Quantity 5 g
Formula Weight 228.09
Percent Purity 98%
Chemical Name or Material 4'-Bromo-3'-methylacetanilide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'