missing translation for 'onlineSavingsMsg'
Learn More

4-bromo-3-methyl-1H-pyrazol-5-amine hydrobromide, Thermo Scientific™

Product Code. 10350673
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10350673 10 g 10g
10692185 1 g 1g
10285284 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10350673 Supplier Thermo Scientific Maybridge Supplier No. KM01090EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 167683-86-5
Molecular Formula C4H7Br2N3
Molecular Weight (g/mol) 256.929
InChI Key UGCOULAJNKWOPX-UHFFFAOYSA-N
Synonym 5-amino-4-bromo-3-methylpyrazole hydrobromide, 4-bromo-3-methyl-1h-pyrazol-5-amine hydrobromide, 4-bromo-5-methyl-1h-pyrazol-3-amine hydrobromide, 3-methyl-4-bromo-5-aminopyrazole hydrobromide, 4-bromo-5-methyl-2h-pyrazol-3-amine hydrobromide, acmc-20am6c, buttpark 48\04-71, 6-amino-3-bromo-2-methylpyrazole hydrobromide, 3-amino-4-bromo-5-methyl-1h-pyrazole hydrobromide, 5-amino-4-bromo-3-methyl-1h-pyrazole hydrobromide
PubChem CID 2778427
IUPAC Name 4-bromo-5-methyl-1H-pyrazol-3-amine;hydrobromide
SMILES CC1=C(C(=NN1)N)Br.Br

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 256.93
Percent Purity 97%
Chemical Name or Material 4-Bromo-3-methyl-1H-pyrazol-5-amine hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.