missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorotoluene, 98%, Thermo Scientific™

Product Code. 10073720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10073720 5 g Glass bottle 5g
10167803 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10073720 Supplier Thermo Scientific Acros Supplier No. 311090050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 452-74-4
MDL Number MFCD00040940
InChI Key SLFNGVGRINFJLK-UHFFFAOYSA-N
Synonym 4-bromo-3-fluorotoluene, 3-fluoro-4-bromotoluene, benzene, 1-bromo-2-fluoro-4-methyl, 3-fluoro-4-bromo-toluene, 4-bromo-3-fluoro-1-methylbenzene, 1-bromo-2-fluoro-4-methyl-benzene, 3-fluoro-4-bromo-toluone, 1-brom-2-fluor-4-methylbenzol, zlchem 562
PubChem CID 573280
IUPAC Name 1-bromo-2-fluoro-4-methylbenzene
SMILES CC1=CC(=C(C=C1)Br)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.4900g/mL
Boiling Point 95.0°C (50.0 mmHg)
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H4BrFO
Linear Formula CH3C6H3(Br)F
Refractive Index 1.5290 to 1.5310
Quantity 5 g
Specific Gravity 1.49
Molecular Weight (g/mol) 191.01
Formula Weight 191.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Bromo-3-fluorotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.