missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluoroaniline, 98%

Product Code. 11465866
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465866 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11465866 Supplier Thermo Scientific Alfa Aesar Supplier No. B22227.09
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 656-65-5 | C6H5BrFN | 190.015 g/mol

4-Bromo-3-fluoroaniline is an intermediate used for the synthetic preparation of Tradizolid and various pharmaceuticals such as the synthesis of several lateral difluoro-substituted 4,4-dialkyl- and 4,4-alkoxyalkylterphenyls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-3-fluoroaniline is an intermediate used for the synthetic preparation of Tradizolid and various pharmaceuticals such as the synthesis of several lateral difluoro-substituted 4,4″-dialkyl- and 4,4″-alkoxyalkylterp
TRUSTED_SUSTAINABILITY

Specifications

CAS 656-65-5
Melting Point 65°C to 70°C
Flash Point 104°C (219°F)
Molecular Formula C6H5BrFN
MDL Number MFCD00672933
Quantity 10 g
UN Number UN2811
Beilstein 3236457
Synonym 4-bromo-3-fluorophenylamine, 3-fluoro-4-bromoaniline, 4-bromo-3-fluoro aniline, benzenamine, 4-bromo-3-fluoro, 4-brom-3-fluoranilin, zlchem 544, pubchem1526, acmc-1badx, 4-bromo-3-fluro-aniline, 4-bromo-3-fluoro-aniline
Solubility Information Soluble in acetonitrile,methanol. Insoluble in water.
InChI Key YTMVYYAKOPIJCZ-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1N)F)Br
IUPAC Name 4-bromo-3-fluoroaniline
Molecular Weight (g/mol) 190.015
PubChem CID 821848
Formula Weight 190.01
Percent Purity 98%
Chemical Name or Material 4-Bromo-3-fluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.