missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-3-chloropropiophenone, 94%

Product Code. 11421758
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
Unit Size:
10g
250g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421758 10 g 10g
11339419 5 g 5g
11431758 50 g 50g
11441758 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11421758 Supplier Thermo Scientific Alfa Aesar Supplier No. A13988.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31736-73-9 | C9H8BrClO | 247.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31736-73-9
Molecular Formula C9H8BrClO
Molecular Weight (g/mol) 247.52
MDL Number MFCD00018979
InChI Key SYWQQEVHZPPLNQ-UHFFFAOYSA-N
Synonym 4'-bromo-3-chloropropiophenone, 1-4-bromophenyl-3-chloropropan-1-one, 4-bromo-3-chloropropiophenone, 1-propanone, 1-4-bromophenyl-3-chloro, pubchem13267, ksc567m6b, 3-chloro-4'-bromopropiophenone, 4'-bromo-beta-chloropropiophenone, 4-3-chloropropanoyl bromobenzene, 4\\'-bromo-3-chloropropiophenone
PubChem CID 95790
IUPAC Name 1-(4-bromophenyl)-3-chloropropan-1-one
SMILES ClCCC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 57°C to 62°C
Quantity 10 g
Beilstein 1944367
Formula Weight 247.53
Percent Purity 94%
Chemical Name or Material 4'-Bromo-3-chloropropiophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.