missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-chlorophenol, 98%, Thermo Scientific Chemicals

Product Code. 11313267
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313267 5 g 5g
11303267 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313267 Supplier Thermo Scientific Alfa Aesar Supplier No. L13801.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 13631-21-5 | Molecular Formula: C6H4BrClO | Molecular Weight: 207.451 g/mol

4-Bromo-3-chlorophenol is used as a precursor in organic synthesis to prepare 4-(benzyloxy)-l-bromo-2-chlorobenzene by reaction with benzyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-3-chlorophenol is used as a precursor in organic synthesis to prepare 4-(benzyloxy)-l-bromo-2-chlorobenzene by reaction with benzyl bromide.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 13631-21-5
Molecular Formula C6H4BrClO
Molecular Weight (g/mol) 207.451
MDL Number MFCD00070744
InChI Key FQEYHIPPYOSPLF-UHFFFAOYSA-N
Synonym phenol, 4-bromo-3-chloro, 3-chloro-4-bromophenol, 2-chloro-4-hydroxybromobenzene, pubchem3597, acmc-1bvpq, 4-bromo-3-chloro-phenol, intermediates-zcf02051, ksc493i5n, 4-bromanyl-3-chloranyl-phenol
PubChem CID 7018182
IUPAC Name 4-bromo-3-chlorophenol
SMILES C1=CC(=C(C=C1O)Cl)Br

Spezifikation

Melting Point 64°C to 68°C
Odor Phenol-like
Quantity 5 g
Beilstein 3236445
Solubility Information Slightly soluble in water.
Formula Weight 207.46
Percent Purity 98%
Chemical Name or Material 4-Bromo-3-chlorophenol

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.