missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-dimethylisoxazole, 97%, Thermo Scientific™

Product Code. 10793147
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10793147 25 g 25g
10254774 1 g 1g
10497174 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10793147 Supplier Thermo Scientific Maybridge Supplier No. CC00610EB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10558-25-5
Molecular Formula C5H6BrNO
Molecular Weight (g/mol) 176.013
MDL Number MFCD00068187
InChI Key GYHZPSUAMYIFQD-UHFFFAOYSA-N
Synonym 4-bromo-3,5-dimethylisoxazole, 3,5-dimethyl-4-bromoisoxazole, isoxazole, 4-bromo-3,5-dimethyl, 4-bromo-dimethylisoxazole, buttpark 33\06-98, 4-bromo-3,5-dimethylisoxazole, tech., pubchem8720, acmc-1bpqs, timtec-bb sbb003784, ksc182i2n
PubChem CID 318421
IUPAC Name 4-bromo-3,5-dimethyl-1,2-oxazole
SMILES CC1=C(C(=NO1)C)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Density 1.484g/mL
Assay Percent Range 97%
Quantity 25 g
Formula Weight 176.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-3, 5-dimethylisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.