Learn More
4-Bromo-3,5-dihydroxybenzoic acid, 98%, Thermo Scientific™
£16.00
Chemical Identifiers
CAS | 16534-12-6 |
---|---|
MDL Number | MFCD00002513 |
InChI Key | NUTRHYYFCDEALP-UHFFFAOYSA-N |
Synonym | benzoic acid, 4-bromo-3,5-dihydroxy, 4-bromo-3,5-resorcylic acid, 4-bromo-alpha-resorcylic acid, 4-bromo-3,5-dihydroxybenzic acid, .alpha.-resorcylic acid, 4-bromo, 4-bromo-3,5-dihydroxy-benzoic acid, alpha-resorcylic acid, 4-bromo, pubchem8196, acmc-209dsh, ksc177s3r |
PubChem CID | 86023 |
IUPAC Name | 4-bromo-3,5-dihydroxybenzoic acid |
SMILES | C1=C(C=C(C(=C1O)Br)O)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10237693
|
Thermo Scientific Acros
394090050 |
5 g | Glass bottle | ||||||
Description
Chemical Identifiers
16534-12-6 | |
NUTRHYYFCDEALP-UHFFFAOYSA-N | |
86023 | |
C1=C(C=C(C(=C1O)Br)O)C(=O)O |
MFCD00002513 | |
benzoic acid, 4-bromo-3,5-dihydroxy, 4-bromo-3,5-resorcylic acid, 4-bromo-alpha-resorcylic acid, 4-bromo-3,5-dihydroxybenzic acid, .alpha.-resorcylic acid, 4-bromo, 4-bromo-3,5-dihydroxy-benzoic acid, alpha-resorcylic acid, 4-bromo, pubchem8196, acmc-209dsh, ksc177s3r | |
4-bromo-3,5-dihydroxybenzoic acid |
Specifications
16534-12-6 | |
100.0 | |
Cream to White or Pink | |
97.5% min. (HPLC) | |
BrC6H2(OH)2CO2H | |
benzoic acid, 4-bromo-3,5-dihydroxy, 4-bromo-3,5-resorcylic acid, 4-bromo-alpha-resorcylic acid, 4-bromo-3,5-dihydroxybenzic acid, .alpha.-resorcylic acid, 4-bromo, 4-bromo-3,5-dihydroxy-benzoic acid, alpha-resorcylic acid, 4-bromo, pubchem8196, acmc-209dsh, ksc177s3r | |
C1=C(C=C(C(=C1O)Br)O)C(=O)O | |
233.02 | |
233.02 | |
Powder |
97.5 | |
269.0°C to 271.0°C | |
Authentic | |
C7H5BrO4 | |
MFCD00002513 | |
NUTRHYYFCDEALP-UHFFFAOYSA-N | |
4-bromo-3,5-dihydroxybenzoic acid | |
86023 | |
98% | |
4-Bromo-3, 5-dihydroxybenzoic acid |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 240-606-4
- 4-Bromo-3,5-dihydroxybenzoic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only